The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024311.3 | NP_077287.1 | 1 - 2 | 386 | 138 |
GenBank mRNA | BC002753.2 | - | 1 - 2 | 186 | 138 |