The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270940.1 | NP_001257869.1 | 17 - 18 | 2841 | 72 |
NM_015171.3 | NP_055986.1 | 16 - 17 | 2705 | 72 | |
XM_005255197.3 | XP_005255254.1 | 14 - 15 | 2039 | 72 | |
XM_005255198.1 | XP_005255255.1 | 12 - 13 | 2188 | 72 | |
XM_005255199.2 | XP_005255256.1 | 12 - 13 | 1382 | 72 | |
GenBank mRNA | AB002368.3 | - | 16 - 17 | 2656 | 72 |
AK026172.1 | - | 13 - 14 | 1849 | 72 | |
AK123846.1 | - | 17 - 18 | 2473 | 72 | |
AY026388.1 | - | 16 - 17 | 2178 | 72 | |
BC004403.1 | - | 10 - 11 | 1445 | 72 | |
BC014071.2 | - | 10 - 11 | 1445 | 72 | |
BC078674.1 | - | 12 - 13 | 1650 | 72 | |
BC108286.1 | - | 12 - 13 | 1604 | 72 | |
BC130304.1 | - | 16 - 17 | 2178 | 72 | |
BC144189.1 | - | 16 - 17 | 2178 | 72 |