Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQGAP1
Entrez Gene ID
8826
Gene Name
IQ motif containing GTPase activating protein 1
Gene Aliases
HUMORFA01, SAR1, p195
Chromosome Location
Chr.15: 90388241 - 90502243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003870.3 NP_003861.1 10 - 11 1178 77
GenBank mRNA AK307643.1 - 10 - 11 1095 77
BC020549.1 - 10 - 11 1122 77
BC037425.1 - 10 - 11 1132 77
BC064971.1 - 10 - 11 1128 77
BC110811.1 - 10 - 11 1113 77
BC139731.1 - 10 - 11 1123 77
BC151834.1 - 10 - 11 1201 77
D29640.1 - 10 - 11 1201 77

View Full Product Details