The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244710.1 | NP_001231639.1 | 13 - 14 | 1392 | 85 |
NM_002056.3 | NP_002047.2 | 12 - 13 | 1338 | 85 | |
XM_017003801.1 | XP_016859290.1 | 13 - 14 | 1356 | 85 | |
XM_017003802.1 | XP_016859291.1 | 12 - 13 | 1294 | 85 | |
GenBank mRNA | BC045641.1 | - | 12 - 13 | 1334 | 85 |
M90516.1 | - | 12 - 13 | 1277 | 85 |