Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFPT1
Entrez Gene ID
2673
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Gene Aliases
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG
Chromosome Location
Chr.2: 69319769 - 69387254 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244710.1 NP_001231639.1 6 - 7 729 79
NM_002056.3 NP_002047.2 6 - 7 729 79
XM_017003801.1 XP_016859290.1 6 - 7 693 79
XM_017003802.1 XP_016859291.1 6 - 7 685 79
GenBank mRNA AF334737.1 - 2 - 3 177 79
BC045641.1 - 6 - 7 725 79
DB133199.1 - 5 - 6 525 80
M90516.1 - 6 - 7 668 79

View Full Product Details