Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFPT1
Entrez Gene ID
2673
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Gene Aliases
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG
Chromosome Location
Chr.2: 69319769 - 69387254 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244710.1 NP_001231639.1 10 - 11 1024 97
NM_002056.3 NP_002047.2 9 - 10 970 97
XM_017003801.1 XP_016859290.1 10 - 11 988 97
XM_017003802.1 XP_016859291.1 9 - 10 926 97
GenBank mRNA BC045641.1 - 9 - 10 966 97
M90516.1 - 9 - 10 909 97

View Full Product Details