The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025083.3 | NP_079359.2 | 1 - 2 | 166 | 91 |
GenBank mRNA | AK056339.1 | - | 1 - 2 | 166 | 91 |
BC011534.1 | - | 1 - 2 | 140 | 91 |