Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
Chromosome Location
Chr.15: 74337762 - 74367740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000781.2 NP_000772.2 1 - 2 424 77
GenBank mRNA AK225715.1 - 1 - 2 330 77
AK292300.1 - 1 - 2 330 77
AK293204.1 - 1 - 2 330 77
AK293211.1 - 1 - 2 330 77
AK300178.1 - 1 - 2 330 77
AK310362.1 - 1 - 2 330 77
BC032329.1 - 1 - 2 313 77
BX398579.2 - 1 - 2 424 77
KU178011.1 - 1 - 2 269 77
M14565.1 - 1 - 2 313 77

View Full Product Details