Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMAN1L
Entrez Gene ID
79748
Gene Name
lectin, mannose binding 1 like
Gene Aliases
ERGIC-53L, ERGL
Chromosome Location
Chr.15: 74812853 - 74825758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021819.2 NP_068591.2 7 - 8 777 78
GenBank mRNA AF303398.1 - 7 - 8 813 78
AK291819.1 - 7 - 8 782 78

View Full Product Details