The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002877.1 | NP_001002877.1 | 18 - 19 | 78 | |
NM_001002878.1 | NP_001002878.1 | 18 - 19 | 78 | ||
NM_001002879.1 | NP_001002879.1 | 17 - 18 | 78 | ||
NM_003678.4 | NP_003669.4 | 17 - 18 | 78 | ||
XM_005261797.1 | XP_005261854.1 | 18 - 19 | 78 | ||
XM_005261798.1 | XP_005261855.1 | 19 - 20 | 78 | ||
XM_005261799.1 | XP_005261856.1 | 18 - 19 | 78 | ||
XM_005261801.3 | XP_005261858.1 | 17 - 18 | 78 | ||
XM_006724347.1 | XP_006724410.1 | 19 - 20 | 78 | ||
XM_017029027.1 | XP_016884516.1 | 19 - 20 | 78 | ||
GenBank mRNA | AB023200.1 | - | 17 - 18 | 78 | |
AJ006069.1 | - | 1 - 2 | 78 | ||
AK025385.1 | - | 17 - 18 | 78 | ||
AK098709.1 | - | 14 - 15 | 78 | ||
AK122673.1 | - | 20 - 21 | 78 | ||
AK225752.1 | - | 16 - 17 | 78 | ||
BC003615.2 | - | 17 - 18 | 78 | ||
BX400943.2 | - | 3 - 4 | 468 | 78 | |
CR456542.1 | - | 16 - 17 | 78 |