The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002877.1 | NP_001002877.1 | 11 - 12 | 1169 | 90 |
NM_001002878.1 | NP_001002878.1 | 11 - 12 | 1300 | 90 | |
NM_001002879.1 | NP_001002879.1 | 10 - 11 | 1184 | 90 | |
NM_003678.4 | NP_003669.4 | 10 - 11 | 1053 | 90 | |
XM_005261797.1 | XP_005261854.1 | 11 - 12 | 1099 | 90 | |
XM_005261798.1 | XP_005261855.1 | 12 - 13 | 1215 | 90 | |
XM_005261799.1 | XP_005261856.1 | 11 - 12 | 1242 | 90 | |
XM_006724347.1 | XP_006724410.1 | 12 - 13 | 1329 | 90 | |
XM_017029027.1 | XP_016884516.1 | 12 - 13 | 1701 | 90 | |
GenBank mRNA | AB023200.1 | - | 10 - 11 | 1020 | 90 |
AK025385.1 | - | 10 - 11 | 1003 | 90 | |
AK098709.1 | - | 7 - 8 | 607 | 90 | |
AK225752.1 | - | 9 - 10 | 786 | 90 | |
BC003615.2 | - | 10 - 11 | 993 | 90 | |
CR456542.1 | - | 9 - 10 | 987 | 90 |