The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002877.1 | NP_001002877.1 | 2 - 3 | 196 | 71 |
NM_001002878.1 | NP_001002878.1 | 2 - 3 | 327 | 71 | |
XM_005261798.1 | XP_005261855.1 | 2 - 3 | 209 | 71 | |
XM_006724347.1 | XP_006724410.1 | 2 - 3 | 323 | 71 | |
XM_017029027.1 | XP_016884516.1 | 2 - 3 | 695 | 71 | |
GenBank mRNA | AK122673.1 | - | 2 - 3 | 703 | 71 |
BX381026.2 | - | 2 - 3 | 161 | 71 | |
CN387648.1 | - | 2 - 3 | 332 | 71 |