Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM39B
Entrez Gene ID
55116
Gene Name
transmembrane protein 39B
Gene Aliases
-
Chromosome Location
Chr.1: 32072031 - 32102866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319677.1 NP_001306606.1 3 - 4 770 84
NM_018056.3 NP_060526.2 3 - 4 501 84
XM_005270988.2 XP_005271045.1 3 - 4 623 84
XM_006710724.1 XP_006710787.1 3 - 4 501 84
XM_006710725.3 XP_006710788.1 3 - 4 501 84
XM_011541681.2 XP_011539983.1 3 - 4 501 84
XM_011541684.2 XP_011539986.1 3 - 4 1369 84
GenBank mRNA AK001177.1 - 3 - 4 472 84
AK023331.1 - 3 - 4 498 84
AK091861.1 - 3 - 4 770 84
AL136695.1 - 3 - 4 479 84
CR457311.1 - 2 - 3 352 84
CR533536.1 - 2 - 3 352 84
DA614559.1 - 3 - 4 501 84

View Full Product Details