Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM39B
Entrez Gene ID
55116
Gene Name
transmembrane protein 39B
Gene Aliases
-
Chromosome Location
Chr.1: 32072031 - 32102866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319677.1 NP_001306606.1 4 - 5 854 67
NM_018056.3 NP_060526.2 4 - 5 585 67
XM_005270988.2 XP_005271045.1 4 - 5 707 67
XM_006710724.1 XP_006710787.1 4 - 5 585 67
XM_006710725.3 XP_006710788.1 4 - 5 585 67
XM_011541681.2 XP_011539983.1 4 - 5 585 67
XM_011541683.1 XP_011539985.1 2 - 3 209 67
XM_017001595.1 XP_016857084.1 2 - 3 331 67
GenBank mRNA AK001177.1 - 4 - 5 556 67
AK023331.1 - 4 - 5 582 67
AK091861.1 - 4 - 5 854 67
AK300293.1 - 5 - 6 571 67
AL136695.1 - 4 - 5 563 67
CR457311.1 - 3 - 4 436 67
CR533536.1 - 3 - 4 436 67

View Full Product Details