The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001931.4 | NP_001922.2 | 1 - 2 | 934 | 76 |
XM_011542647.2 | XP_011540949.1 | 1 - 2 | 581 | 76 | |
GenBank mRNA | AK057299.1 | - | 1 - 2 | 934 | 76 |
AK223596.1 | - | 1 - 2 | 323 | 76 | |
AK297103.1 | - | 1 - 2 | 337 | 76 | |
AK299562.1 | - | 1 - 2 | 152 | 76 | |
AK303930.1 | - | 1 - 2 | 340 | 76 | |
AL832703.1 | - | 1 - 2 | 306 | 76 | |
BC039084.1 | - | 1 - 2 | 303 | 76 | |
Y00978.1 | - | 1 - 2 | 179 | 76 |