The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001931.4 | NP_001922.2 | 2 - 3 | 1037 | 103 |
XM_011542647.2 | XP_011540949.1 | 2 - 3 | 684 | 103 | |
GenBank mRNA | AK057299.1 | - | 2 - 3 | 1037 | 103 |
AK223596.1 | - | 2 - 3 | 426 | 103 | |
AK296274.1 | - | 2 - 3 | 316 | 103 | |
AL832703.1 | - | 2 - 3 | 408 | 102 | |
BC039084.1 | - | 2 - 3 | 406 | 103 |