Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL11A2
Entrez Gene ID
1302
Gene Name
collagen type XI alpha 2
Gene Aliases
DFNA13, DFNB53, FBCG2, HKE5, PARP, STL3
Chromosome Location
Chr.6: 33162692 - 33193009 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080679.2 NP_542410.2 27 - 28 2282 135
NM_080680.2 NP_542411.2 30 - 31 2603 135
NM_080681.2 NP_542412.2 28 - 29 2345 135
XM_011514299.2 XP_011512601.1 25 - 26 1894 135
XM_011514300.2 XP_011512602.1 24 - 25 1714 135
XM_011514302.2 XP_011512604.1 23 - 24 1915 135
XM_017010250.1 XP_016865739.1 31 - 32 2868 135
XM_017010251.1 XP_016865740.1 23 - 24 1732 135
GenBank mRNA J04974.1 - 3 - 4 188 135
L18987.1 - 27 - 28 1964 135

View Full Product Details