The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080679.2 | NP_542410.2 | 51 - 52 | 3865 | 118 |
NM_080680.2 | NP_542411.2 | 54 - 55 | 4186 | 118 | |
NM_080681.2 | NP_542412.2 | 52 - 53 | 3928 | 118 | |
XM_011514299.2 | XP_011512601.1 | 49 - 50 | 3477 | 118 | |
XM_011514300.2 | XP_011512602.1 | 48 - 49 | 3297 | 118 | |
XM_011514302.2 | XP_011512604.1 | 47 - 48 | 3498 | 118 | |
XM_017010250.1 | XP_016865739.1 | 55 - 56 | 4451 | 118 | |
XM_017010251.1 | XP_016865740.1 | 47 - 48 | 3315 | 118 | |
GenBank mRNA | J04974.1 | - | 27 - 28 | 1771 | 118 |