Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
Chromosome Location
Chr.15: 89627970 - 89663086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198525.2 NP_940927.2 12 - 13 2672 76
XM_011521531.2 XP_011519833.1 12 - 13 2782 76
GenBank mRNA AY358384.1 - 8 - 9 1241 76
BC104044.1 - 8 - 9 1214 76
BC112271.1 - 7 - 8 1105 76
BC112273.1 - 7 - 8 1105 76
BC143461.1 - 7 - 8 1101 76

View Full Product Details