Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
Chromosome Location
Chr.15: 89627970 - 89663086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198525.2 NP_940927.2 14 - 15 2972 93
XM_011521531.2 XP_011519833.1 14 - 15 3082 93
GenBank mRNA AY358384.1 - 10 - 11 1541 93
BC040878.1 - 2 - 3 271 93
BC104044.1 - 10 - 11 1514 93
BC112271.1 - 9 - 10 1405 93
BC112273.1 - 9 - 10 1405 93
BC143461.1 - 9 - 10 1401 93

View Full Product Details