The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021819.2 | NP_068591.2 | 6 - 7 | 721 | 79 |
GenBank mRNA | AF303398.1 | - | 6 - 7 | 757 | 79 |
AK291819.1 | - | 6 - 7 | 726 | 79 |