Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UQCRC1
Entrez Gene ID
7384
Gene Name
ubiquinol-cytochrome c reductase core protein I
Gene Aliases
D3S3191, QCR1, UQCR1
Chromosome Location
Chr.3: 48598999 - 48609665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003365.2 NP_003356.2 1 - 2 116 112
GenBank mRNA AK129903.1 - 1 - 2 96 112
AK313090.1 - 1 - 2 86 112
BC009586.1 - 1 - 2 87 112
D26485.1 - 1 - 2 78 112
L16842.1 - 1 - 2 488 112

View Full Product Details