The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320033.1 | NP_001306962.1 | 7 - 8 | 1206 | 92 |
NM_004803.4 | NP_004794.2 | 6 - 7 | 1251 | 92 | |
XM_005265585.4 | XP_005265642.1 | 7 - 8 | 2028 | 92 | |
XM_006713416.3 | XP_006713479.2 | 7 - 8 | 1952 | 92 | |
XM_006713418.3 | XP_006713481.2 | 7 - 8 | 1952 | 92 | |
XM_011534245.2 | XP_011532547.1 | 7 - 8 | 2040 | 92 | |
GenBank mRNA | AB011082.1 | - | 6 - 7 | 1250 | 92 |
AK315317.1 | - | 7 - 8 | 1204 | 92 | |
BC075070.2 | - | 6 - 7 | 1195 | 92 | |
BC075071.2 | - | 6 - 7 | 1195 | 92 | |
BC126482.1 | - | 6 - 7 | 1232 | 92 | |
BC130543.1 | - | 6 - 7 | 1232 | 92 | |
BC144079.1 | - | 6 - 7 | 1191 | 92 |