The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278738.1 | NP_001265667.1 | 8 - 9 | 1251 | 90 |
NM_001278739.1 | NP_001265668.1 | 7 - 8 | 1136 | 90 | |
NM_016593.4 | NP_057677.2 | 8 - 9 | 1311 | 90 | |
XM_005249171.3 | XP_005249228.1 | 8 - 9 | 1286 | 90 | |
XM_017010921.1 | XP_016866410.1 | 8 - 9 | 1286 | 90 | |
XM_017010922.1 | XP_016866411.1 | 8 - 9 | 1270 | 90 | |
XM_017010923.1 | XP_016866412.1 | 7 - 8 | 1178 | 90 | |
XM_017010924.1 | XP_016866413.1 | 7 - 8 | 1178 | 90 | |
GenBank mRNA | AF237982.1 | - | 8 - 9 | 1107 | 90 |
AK292263.1 | - | 8 - 9 | 1243 | 90 | |
AK301604.1 | - | 8 - 9 | 1129 | 90 | |
AK307986.1 | - | 7 - 8 | 800 | 90 | |
BC010358.1 | - | 8 - 9 | 1271 | 90 |