The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006270.4 | NP_006261.1 | 3 - 4 | 396 | 81 |
GenBank mRNA | AF493920.1 | - | 3 - 4 | 351 | 81 |
BC016286.1 | - | 3 - 4 | 375 | 81 | |
BC016318.1 | - | 3 - 4 | 400 | 81 | |
BT006805.1 | - | 3 - 4 | 351 | 81 | |
CR541944.1 | - | 3 - 4 | 351 | 81 | |
CR541967.1 | - | 3 - 4 | 351 | 81 |