The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024094.2 | NP_076999.2 | 4 - 5 | 710 | 140 |
XM_005251065.3 | XP_005251122.1 | 3 - 4 | 566 | 140 | |
GenBank mRNA | AK054585.1 | - | 4 - 5 | 682 | 140 |
AK074355.1 | - | 4 - 5 | 647 | 140 | |
AK315356.1 | - | 4 - 5 | 636 | 140 | |
AY358866.1 | - | 4 - 5 | 662 | 140 | |
BC001316.1 | - | 4 - 5 | 689 | 140 | |
BC001531.2 | - | 4 - 5 | 648 | 140 |