The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024094.2 | NP_076999.2 | 8 - 9 | 1209 | 77 |
XM_005251065.3 | XP_005251122.1 | 7 - 8 | 1065 | 77 | |
GenBank mRNA | AK054585.1 | - | 8 - 9 | 1181 | 77 |
AK074355.1 | - | 8 - 9 | 1146 | 77 | |
AK315356.1 | - | 8 - 9 | 1135 | 77 | |
AY358866.1 | - | 8 - 9 | 1161 | 77 | |
BC001316.1 | - | 8 - 9 | 1188 | 77 | |
BC001531.2 | - | 8 - 9 | 1147 | 77 |