The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001275.3 | NP_001266.1 | 2 - 3 | 354 | 67 |
NM_001301690.1 | NP_001288619.1 | 2 - 3 | 354 | 67 | |
XM_011536370.1 | XP_011534672.1 | 3 - 4 | 706 | 67 | |
GenBank mRNA | AK223381.1 | - | 2 - 3 | 301 | 67 |
AK313757.1 | - | 2 - 3 | 305 | 67 | |
BC001059.2 | - | 2 - 3 | 154 | 67 | |
BC006459.1 | - | 2 - 3 | 294 | 67 | |
BT006869.1 | - | 2 - 3 | 94 | 67 | |
BX248263.1 | - | 2 - 3 | 278 | 67 | |
DA201149.1 | - | 2 - 3 | 354 | 67 | |
J03483.1 | - | 2 - 3 | 176 | 67 |