Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAVER2
Entrez Gene ID
55225
Gene Name
ribonucleoprotein, PTB binding 2
Gene Aliases
-
Chromosome Location
Chr.1: 64744979 - 64833232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018211.3 NP_060681.2 11 - 12 1970 80
XM_006710738.3 XP_006710801.2 11 - 12 2116 80
GenBank mRNA AB046799.1 - 11 - 12 1970 80
AK299228.1 - 6 - 7 679 80
AL359613.1 - 4 - 5 576 80
BC065303.1 - 9 - 10 1400 80

View Full Product Details