The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018211.3 | NP_060681.2 | 8 - 9 | 1453 | 67 |
XM_006710738.3 | XP_006710801.2 | 8 - 9 | 1599 | 67 | |
GenBank mRNA | AB046799.1 | - | 8 - 9 | 1453 | 67 |
AL359613.1 | - | 1 - 2 | 59 | 67 | |
BC065303.1 | - | 6 - 7 | 883 | 67 |