Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAVER2
Entrez Gene ID
55225
Gene Name
ribonucleoprotein, PTB binding 2
Gene Aliases
-
Chromosome Location
Chr.1: 64744979 - 64833232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018211.3 NP_060681.2 9 - 10 1721 69
XM_006710738.3 XP_006710801.2 9 - 10 1867 69
GenBank mRNA AB046799.1 - 9 - 10 1721 69
AL359613.1 - 2 - 3 327 69
BC065303.1 - 7 - 8 1151 69

View Full Product Details