The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153603.3 | NP_705831.1 | 6 - 7 | 1014 | 77 |
XM_017023870.1 | XP_016879359.1 | 6 - 7 | 1057 | 77 | |
GenBank mRNA | AK093989.1 | - | 3 - 4 | 370 | 77 |
AY358632.1 | - | 1 - 2 | 51 | 77 | |
BC000549.2 | - | 6 - 7 | 969 | 77 | |
BC037563.1 | - | 6 - 7 | 1003 | 77 | |
KU178837.1 | - | 6 - 7 | 817 | 77 |