The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022351.4 | NP_071746.1 | 5 - 6 | 554 | 98 |
XM_011517213.2 | XP_011515515.1 | 5 - 6 | 479 | 98 | |
GenBank mRNA | AF193756.1 | - | 1 - 2 | 72 | 98 |
AF414126.2 | - | 6 - 7 | 643 | 98 | |
AK095533.1 | - | 5 - 6 | 467 | 98 | |
AY224210.1 | - | 5 - 6 | 360 | 98 | |
BC068448.1 | - | 5 - 6 | 467 | 98 |