The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013353.2 | NP_037485.2 | 2 - 3 | 307 | 78 |
XM_011509449.1 | XP_011507751.1 | 2 - 3 | 189 | 78 | |
XM_017001089.1 | XP_016856578.1 | 1 - 2 | 924 | 78 | |
XM_017001090.1 | XP_016856579.1 | 2 - 3 | 236 | 78 | |
GenBank mRNA | AF165217.1 | - | 2 - 3 | 184 | 78 |
AF177173.2 | - | 2 - 3 | 252 | 78 | |
AK300675.1 | - | 2 - 3 | 217 | 78 | |
AK313280.1 | - | 2 - 3 | 216 | 78 | |
BC017810.1 | - | 2 - 3 | 217 | 78 | |
BX647346.1 | - | 2 - 3 | 217 | 78 | |
DC385872.1 | - | 2 - 3 | 307 | 78 |