Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMOD4
Entrez Gene ID
29765
Gene Name
tropomodulin 4
Gene Aliases
SK-TMOD
Chromosome Location
Chr.1: 151169986 - 151176071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013353.2 NP_037485.2 8 - 9 1063 65
XM_011509449.1 XP_011507751.1 8 - 9 945 65
XM_017001089.1 XP_016856578.1 7 - 8 1680 65
GenBank mRNA AF177173.2 - 8 - 9 1008 65
AK092700.1 - 8 - 9 925 65
AK300675.1 - 6 - 7 766 65
AK313280.1 - 8 - 9 972 65
BC017810.1 - 8 - 9 973 65
BX647346.1 - 6 - 7 687 65

View Full Product Details