Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBNL
Entrez Gene ID
28988
Gene Name
drebrin like
Gene Aliases
ABP1, HIP-55, HIP55, SH3P7
Chromosome Location
Chr.7: 44044640 - 44061716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014436.2 NP_001014436.1 7 - 8 805 84
NM_001284313.1 NP_001271242.1 5 - 6 636 84
XM_011515325.1 XP_011513627.1 7 - 8 776 84
GenBank mRNA AB209486.1 - 5 - 6 492 84
AF077353.1 - 7 - 8 745 84
AF197060.1 - 7 - 8 707 84
AK126142.1 - 4 - 5 364 84
AK293340.1 - 6 - 7 624 84
AK293698.1 - 5 - 6 601 84
AK296778.1 - 6 - 7 557 84
AK298102.1 - 6 - 7 557 84
BC031687.1 - 7 - 8 713 84
CR457167.1 - 7 - 8 707 84

View Full Product Details