The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178074.1 | NP_001171545.1 | 7 - 8 | 964 | 117 |
NM_021150.3 | NP_066973.2 | 7 - 8 | 964 | 117 | |
XM_005268754.4 | XP_005268811.1 | 7 - 8 | 884 | 117 | |
XM_005268757.4 | XP_005268814.1 | 6 - 7 | 803 | 117 | |
XM_011538093.2 | XP_011536395.1 | 7 - 8 | 937 | 117 | |
XM_017019098.1 | XP_016874587.1 | 7 - 8 | 1233 | 117 | |
XM_017019099.1 | XP_016874588.1 | 7 - 8 | 3892 | 117 | |
XM_017019100.1 | XP_016874589.1 | 7 - 8 | 1233 | 117 | |
XM_017019101.1 | XP_016874590.1 | 7 - 8 | 3892 | 117 | |
XM_017019102.1 | XP_016874591.1 | 7 - 8 | 3892 | 117 | |
GenBank mRNA | AK095519.1 | - | 7 - 8 | 964 | 117 |
AK295565.1 | - | 6 - 7 | 770 | 117 | |
AK307576.1 | - | 7 - 8 | 802 | 117 | |
BC029871.1 | - | 6 - 7 | 684 | 117 | |
BC115393.1 | - | 7 - 8 | 792 | 117 | |
BC115394.1 | - | 7 - 8 | 792 | 117 |