The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004531.4 | NP_004522.1 | 5 - 6 | 1068 | 85 |
NM_176806.3 | NP_789776.1 | 5 - 6 | 607 | 85 | |
GenBank mRNA | AF091871.1 | - | 5 - 6 | 580 | 85 |
AF117815.1 | - | 5 - 6 | 591 | 85 | |
AF155659.1 | - | 5 - 6 | 793 | 85 | |
AK312887.1 | - | 5 - 6 | 593 | 85 | |
BC029287.1 | - | 5 - 6 | 570 | 85 | |
BC039720.2 | - | 4 - 5 | 548 | 85 | |
BC046097.1 | - | 5 - 6 | 754 | 85 | |
BC095417.1 | - | 5 - 6 | 608 | 85 | |
CD514049.1 | - | 5 - 6 | 603 | 85 | |
CR457172.1 | - | 3 - 4 | 377 | 85 |