Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Chromosome Location
Chr.1: 157676481 - 157700985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320333.1 NP_001307262.1 13 - 14 2331 128
NM_052939.3 NP_443171.2 13 - 14 2316 128
NR_135214.1 13 - 14 2349 128
NR_135215.1 13 - 14 2331 128
NR_135216.1 13 - 14 2236 128
NR_135217.1 12 - 13 2046 128
XM_006711145.1 XP_006711208.1 13 - 14 2246 128
GenBank mRNA AF416901.1 - 13 - 14 2195 128
AF416902.1 - 12 - 13 1910 128
AF416903.1 - 13 - 14 2213 128
AF416904.1 - 12 - 13 2429 128
AF416905.1 - 13 - 14 2100 128
AF459027.1 - 13 - 14 2316 128
AK098122.1 - 13 - 14 2246 128
AK125622.1 - 2 - 3 149 128
AL833391.1 - 7 - 8 2866 128
BC028933.1 - 13 - 14 2197 128

View Full Product Details