Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Chromosome Location
Chr.1: 157676481 - 157700985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320333.1 NP_001307262.1 9 - 10 1993 64
NM_052939.3 NP_443171.2 9 - 10 1978 64
NR_135214.1 9 - 10 2011 64
NR_135215.1 9 - 10 1993 64
NR_135216.1 9 - 10 1898 64
NR_135217.1 8 - 9 1708 64
XM_006711145.1 XP_006711208.1 9 - 10 1908 64
GenBank mRNA AF416901.1 - 9 - 10 1857 64
AF416902.1 - 8 - 9 1572 64
AF416903.1 - 9 - 10 1875 64
AF416904.1 - 8 - 9 2091 64
AF416905.1 - 9 - 10 1762 64
AF459027.1 - 9 - 10 1978 64
AK098122.1 - 9 - 10 1908 64
AK301257.1 - 8 - 9 2114 64
AL833391.1 - 3 - 4 2528 64
AY043466.1 - 8 - 9 1782 64
BC028933.1 - 9 - 10 1859 64

View Full Product Details