The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297655.1 | NP_001284584.1 | 10 - 11 | 1067 | 67 |
NM_001297656.1 | NP_001284585.1 | 10 - 11 | 1308 | 67 | |
NM_001297657.1 | NP_001284586.1 | 10 - 11 | 995 | 67 | |
NM_006845.3 | NP_006836.2 | 11 - 12 | 1190 | 67 | |
XM_011540540.2 | XP_011538842.1 | 11 - 12 | 1437 | 67 | |
XM_011540541.2 | XP_011538843.1 | 10 - 11 | 1378 | 67 | |
GenBank mRNA | AB264115.1 | - | 11 - 12 | 1190 | 67 |
AK290688.1 | - | 11 - 12 | 1116 | 67 | |
AK300756.1 | - | 10 - 11 | 1024 | 67 | |
AK302267.1 | - | 10 - 11 | 963 | 67 | |
AK312606.1 | - | 11 - 12 | 1146 | 67 | |
AY026505.1 | - | 10 - 11 | 1261 | 67 | |
BC008764.2 | - | 11 - 12 | 1129 | 67 | |
BC014924.1 | - | 11 - 12 | 1127 | 67 | |
BC032038.1 | - | 3 - 4 | 305 | 67 | |
BT006759.1 | - | 11 - 12 | 1075 | 67 | |
CR450302.1 | - | 11 - 12 | 1075 | 67 | |
U63743.1 | - | 11 - 12 | 1129 | 67 |