The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297655.1 | NP_001284584.1 | 15 - 16 | 1678 | 74 |
NM_001297656.1 | NP_001284585.1 | 15 - 16 | 1919 | 74 | |
NM_001297657.1 | NP_001284586.1 | 15 - 16 | 1606 | 74 | |
NM_006845.3 | NP_006836.2 | 16 - 17 | 1801 | 74 | |
XM_011540540.2 | XP_011538842.1 | 16 - 17 | 2048 | 74 | |
XM_011540541.2 | XP_011538843.1 | 15 - 16 | 1989 | 74 | |
GenBank mRNA | AB264115.1 | - | 16 - 17 | 1801 | 74 |
AK290688.1 | - | 16 - 17 | 1727 | 74 | |
AK300756.1 | - | 15 - 16 | 1635 | 74 | |
AK302267.1 | - | 15 - 16 | 1574 | 74 | |
AK312606.1 | - | 16 - 17 | 1757 | 74 | |
AY026505.1 | - | 15 - 16 | 1872 | 74 | |
BC008764.2 | - | 16 - 17 | 1740 | 74 | |
BC014924.1 | - | 16 - 17 | 1738 | 74 | |
BC032038.1 | - | 8 - 9 | 916 | 74 | |
BT006759.1 | - | 16 - 17 | 1686 | 74 | |
CR450302.1 | - | 16 - 17 | 1686 | 74 | |
U63743.1 | - | 16 - 17 | 1740 | 74 |