Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
Chromosome Location
Chr.8: 25458569 - 25511308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317906.1 NP_001304835.1 10 - 11 2074 171
NM_001317907.1 NP_001304836.1 11 - 12 1991 171
NM_152562.3 NP_689775.2 11 - 12 1991 171
XM_006716294.2 XP_006716357.1 11 - 12 2006 171
XM_011544415.1 XP_011542717.1 12 - 13 2078 171
XM_011544416.2 XP_011542718.1 12 - 13 1772 171
XM_011544417.2 XP_011542719.1 12 - 13 1829 171
XM_011544418.1 XP_011542720.1 12 - 13 2042 171
GenBank mRNA AK092164.1 - 6 - 7 608 171
AK292505.1 - 10 - 11 1806 171
AK299278.1 - 11 - 12 1697 171
AL833396.1 - 8 - 9 1096 171
AL833627.1 - 10 - 11 1768 171
BC036214.1 - 8 - 9 1119 171
BX649008.1 - 7 - 8 982 171

View Full Product Details