Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
Chromosome Location
Chr.8: 25458569 - 25511308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317906.1 NP_001304835.1 6 - 7 1447 66
NM_001317907.1 NP_001304836.1 7 - 8 1364 66
NM_152562.3 NP_689775.2 7 - 8 1364 66
XM_006716294.2 XP_006716357.1 7 - 8 1379 66
GenBank mRNA AK292505.1 - 6 - 7 1179 66
AK299278.1 - 7 - 8 1070 66
AL833396.1 - 4 - 5 469 66
AL833627.1 - 6 - 7 1141 66
BC036214.1 - 4 - 5 492 66

View Full Product Details