The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127889.4 | NP_001121361.1 | 5 - 6 | 575 | 74 |
NM_001127890.4 | NP_001121362.1 | 4 - 5 | 675 | 74 | |
NM_016630.6 | NP_057714.1 | 5 - 6 | 756 | 74 | |
XM_005254437.4 | XP_005254494.1 | 6 - 7 | 789 | 74 | |
XM_017022297.1 | XP_016877786.1 | 5 - 6 | 1124 | 74 | |
XM_017022298.1 | XP_016877787.1 | 6 - 7 | 1214 | 74 | |
GenBank mRNA | AF208861.1 | - | 4 - 5 | 471 | 74 |
AF212231.1 | - | 5 - 6 | 543 | 74 | |
AK123725.1 | - | 3 - 4 | 988 | 74 | |
AK172849.1 | - | 5 - 6 | 588 | 74 | |
AK225527.1 | - | 5 - 6 | 588 | 74 | |
AK301362.1 | - | 4 - 5 | 640 | 74 | |
AK312943.1 | - | 5 - 6 | 590 | 74 | |
BC000244.1 | - | 5 - 6 | 626 | 74 | |
BI771887.1 | - | 4 - 5 | 509 | 74 | |
BX446863.2 | - | 5 - 6 | 706 | 74 |