The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033393.2 | NP_203751.2 | 1 - 2 | 565 | 108 |
XM_005263319.3 | XP_005263376.1 | 2 - 3 | 706 | 108 | |
XM_006714393.3 | XP_006714456.1 | 2 - 3 | 715 | 108 | |
XM_011532388.1 | XP_011530690.1 | 4 - 5 | 1197 | 108 | |
XM_011532389.1 | XP_011530691.1 | 2 - 3 | 1311 | 108 | |
GenBank mRNA | AB051514.1 | - | 1 - 2 | 452 | 108 |