The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033393.2 | NP_203751.2 | 6 - 7 | 997 | 86 |
XM_005263319.3 | XP_005263376.1 | 7 - 8 | 1138 | 86 | |
XM_006714393.3 | XP_006714456.1 | 7 - 8 | 1147 | 86 | |
XM_011532388.1 | XP_011530690.1 | 9 - 10 | 1629 | 86 | |
XM_011532389.1 | XP_011530691.1 | 7 - 8 | 1743 | 86 | |
XM_017008787.1 | XP_016864276.1 | 5 - 6 | 982 | 86 | |
GenBank mRNA | AB051514.1 | - | 6 - 7 | 884 | 86 |