Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
Chromosome Location
Chr.2: 61177418 - 61191210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321300.1 NP_001308229.1 3 - 4 1242 106
NM_001321303.1 NP_001308232.1 2 - 3 1838 106
NM_001321304.1 NP_001308233.1 2 - 3 1838 106
NM_001321305.1 NP_001308234.1 2 - 3 1838 106
NM_152392.4 NP_689605.1 2 - 3 1838 106
GenBank mRNA AK092505.1 - 2 - 3 225 106
AK092817.1 - 2 - 3 1242 106
AL833813.1 - 2 - 3 1573 106
BC063789.1 - 2 - 3 1727 106
BC113671.1 - 2 - 3 835 106
BC117278.1 - 2 - 3 835 106

View Full Product Details