The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024652.4 | NP_078928.3 | 29 - 30 | 5038 | 85 |
XM_005254979.3 | XP_005255036.1 | 15 - 16 | 2696 | 85 | |
XM_011522012.2 | XP_011520314.1 | 31 - 32 | 5312 | 85 | |
XM_011522013.2 | XP_011520315.1 | 29 - 30 | 5021 | 85 | |
XM_011522014.2 | XP_011520316.1 | 29 - 30 | 5236 | 85 | |
XM_011522015.2 | XP_011520317.1 | 24 - 25 | 5831 | 85 | |
XM_011522016.2 | XP_011520318.1 | 27 - 28 | 4513 | 85 | |
XM_011522017.2 | XP_011520319.1 | 15 - 16 | 2703 | 85 | |
XM_011522018.2 | XP_011520320.1 | 15 - 16 | 2698 | 85 | |
XM_017022570.1 | XP_016878059.1 | 19 - 20 | 3285 | 85 | |
GenBank mRNA | AB058693.1 | - | 15 - 16 | 2743 | 85 |
AK054662.1 | - | 3 - 4 | 486 | 85 | |
AK058028.1 | - | 9 - 10 | 1586 | 85 | |
AK131075.1 | - | 10 - 11 | 1971 | 85 | |
BC005408.1 | - | 4 - 5 | 709 | 85 | |
BC020379.1 | - | 4 - 5 | 709 | 85 | |
BC144705.1 | - | 27 - 28 | 4426 | 85 | |
BC146769.1 | - | 15 - 16 | 2743 | 85 | |
BC171802.1 | - | 28 - 29 | 4712 | 85 | |
DQ013130.1 | - | 28 - 29 | 4676 | 85 |