The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024652.4 | NP_078928.3 | 15 - 16 | 2427 | 88 |
XM_011522012.2 | XP_011520314.1 | 17 - 18 | 2701 | 88 | |
XM_011522013.2 | XP_011520315.1 | 15 - 16 | 2410 | 88 | |
XM_011522014.2 | XP_011520316.1 | 15 - 16 | 2625 | 88 | |
XM_011522015.2 | XP_011520317.1 | 10 - 11 | 3220 | 88 | |
XM_011522016.2 | XP_011520318.1 | 13 - 14 | 1902 | 88 | |
XM_011522019.2 | XP_011520321.1 | 15 - 16 | 2625 | 88 | |
XM_017022570.1 | XP_016878059.1 | 5 - 6 | 674 | 88 | |
GenBank mRNA | AB058693.1 | - | 1 - 2 | 88 | |
BC146769.1 | - | 1 - 2 | 88 | ||
DQ013130.1 | - | 14 - 15 | 88 |